Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267025.1 | NP_001253954.1 | 197 | 155 | ||
GenBank mRNA | JU475196.1 | - | 197 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |