Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194122.2 | NP_001181051.1 | 634 | 75 | ||
XM_015124981.2 | XP_014980467.2 | 720 | 75 | |||
XM_015124982.2 | XP_014980468.2 | 1054 | 75 | |||
XM_015124984.2 | XP_014980470.1 | 1005 | 75 | |||
GenBank mRNA | JU320188.1 | - | 627 | 75 | ||
JU320189.1 | - | 626 | 75 | |||
JV045506.1 | - | 628 | 75 | |||
JV489308.1 | - | 631 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |