Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261123.1 | NP_001248052.1 | 373 | 122 | ||
XM_015115289.2 | XP_014970775.1 | 638 | 122 | |||
XM_015115290.2 | XP_014970776.1 | 762 | 122 | |||
XM_028832595.1 | XP_028688428.1 | 1080 | 122 | |||
GenBank mRNA | JU334030.1 | - | 471 | 122 | ||
JV046779.1 | - | 373 | 122 | |||
JV648511.1 | - | 380 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |