Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261372.1 | NP_001248301.1 | 463 | 117 | ||
GenBank mRNA | JU325341.1 | - | 389 | 117 | ||
JU471688.1 | - | 463 | 117 | |||
JV058195.1 | - | 365 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |