Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257734.1 | NP_001244663.1 | 1951 | 106 | ||
XM_015131520.2 | XP_014987006.1 | 2622 | 106 | |||
XM_015131521.2 | XP_014987007.1 | 2714 | 106 | |||
XM_015131524.2 | XP_014987010.1 | 2709 | 106 | |||
XM_015131526.2 | XP_014987012.1 | 2290 | 106 | |||
XM_015131527.2 | XP_014987013.1 | 2596 | 106 | |||
XM_028843069.1 | XP_028698902.1 | 2683 | 106 | |||
XM_028843070.1 | XP_028698903.1 | 2683 | 106 | |||
XM_028843071.1 | XP_028698904.1 | 3355 | 106 | |||
XM_028843072.1 | XP_028698905.1 | 3424 | 106 | |||
XM_028843073.1 | XP_028698906.1 | 3287 | 106 | |||
XM_028843074.1 | XP_028698907.1 | 3287 | 106 | |||
XM_028843075.1 | XP_028698908.1 | 2931 | 106 | |||
XM_028843076.1 | XP_028698909.1 | 2656 | 106 | |||
XM_028843077.1 | XP_028698910.1 | 2783 | 106 | |||
XM_028843078.1 | XP_028698911.1 | 2682 | 106 | |||
XM_028843079.1 | XP_028698912.1 | 2263 | 106 | |||
GenBank mRNA | JU321927.1 | - | 1951 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |