Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001115116.4 | XP_001115116.1 | 1612 | 133 | ||
XM_028828265.1 | XP_028684098.1 | 1523 | 133 | |||
GenBank mRNA | JU472404.1 | - | 1595 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |