Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115619.2 | XP_014971105.2 | 318 | 98 | ||
XM_015115621.2 | XP_014971107.2 | 318 | 98 | |||
XM_015115622.2 | XP_014971108.2 | 318 | 98 |