Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194681.2 | NP_001181610.2 | 555 | 98 |
Gene Symbol: LOC114675467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842710.1 | XP_028698543.1 | 554 | 98 |