Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194681.2 | NP_001181610.2 | 785 | 69 |
Gene Symbol: LOC114675467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842710.1 | XP_028698543.1 | 778 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |