Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086428.4 | XP_001086428.3 | 6616 | 104 | ||
GenBank mRNA | JU484019.1 | - | 1639 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |