Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260587.2 | NP_001247516.1 | 343 | 70 | ||
XM_015121729.2 | XP_014977215.1 | 302 | 70 | |||
XM_015121733.2 | XP_014977219.1 | 533 | 70 | |||
XM_015121734.2 | XP_014977220.1 | 533 | 70 | |||
XM_015121735.2 | XP_014977221.1 | 303 | 70 | |||
GenBank mRNA | JU321611.1 | - | 290 | 70 | ||
JU475069.1 | - | 342 | 70 | |||
JV046690.1 | - | 344 | 70 | |||
JV612369.1 | - | 357 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |