Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260587.2 | NP_001247516.1 | 508 | 82 | ||
XM_015121730.2 | XP_014977216.1 | 568 | 82 | |||
XM_015121731.2 | XP_014977217.1 | 445 | 82 | |||
XM_015121733.2 | XP_014977219.1 | 698 | 82 | |||
XM_015121734.2 | XP_014977220.1 | 698 | 82 | |||
XM_015121735.2 | XP_014977221.1 | 468 | 82 | |||
GenBank mRNA | DQ148062.1 | - | 253 | 82 | ||
JU321611.1 | - | 455 | 82 | |||
JU475069.1 | - | 507 | 82 | |||
JV046690.1 | - | 509 | 82 | |||
JV612369.1 | - | 522 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |