Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258114.1 | NP_001245043.1 | 555 | 124 | ||
XM_015144016.2 | XP_014999502.1 | 744 | 124 | |||
XM_015144017.2 | XP_014999503.1 | 721 | 124 | |||
XM_015144018.1 | XP_014999504.1 | 467 | 124 | |||
GenBank mRNA | JU475108.1 | - | 555 | 124 | ||
JV636092.1 | - | 610 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |