Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130085.2 | XP_014985571.2 | 1462 | 71 | ||
XM_028844536.1 | XP_028700369.1 | 1462 | 71 | |||
GenBank mRNA | JU325719.1 | - | 878 | 71 | ||
JV062333.1 | - | 1308 | 71 | |||
JV508396.1 | - | 1748 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |