Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266433.1 | NP_001253362.1 | 233 | 153 | ||
GenBank mRNA | JU334733.1 | - | 276 | 153 | ||
JU475784.1 | - | 233 | 153 | |||
JV470133.1 | - | 257 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |