Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSUH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097726.4 | XP_001097726.3 | 1326 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |