Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111564.2 | XP_014967050.1 | 1695 | 104 | ||
XM_015111565.2 | XP_014967051.1 | 293 | 104 | |||
GenBank mRNA | JU334773.1 | - | 1450 | 104 | ||
JU475814.1 | - | 1425 | 104 | |||
JV047240.1 | - | 1438 | 104 | |||
JV716250.1 | - | 246 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |