Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257619.1 | NP_001244548.1 | 904 | 158 | ||
GenBank mRNA | JU334874.1 | - | 904 | 158 | ||
JU475893.1 | - | 927 | 158 | |||
JV045732.1 | - | 932 | 158 | |||
JV653292.1 | - | 909 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |