Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266488.1 | NP_001253417.1 | 300 | 141 | ||
GenBank mRNA | JU475957.1 | - | 313 | 141 | ||
JV045759.1 | - | 300 | 141 | |||
JV584240.1 | - | 321 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |