Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145102.2 | XP_015000588.1 | 900 | 141 | ||
XM_028852526.1 | XP_028708359.1 | 533 | 141 | |||
GenBank mRNA | JU322457.1 | - | 420 | 141 | ||
JU475967.1 | - | 422 | 141 | |||
JV045475.1 | - | 313 | 141 | |||
JV638187.1 | - | 418 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |