Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETV7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001116763.4 | XP_001116763.4 | 1519 | 145 | ||
XM_028847620.1 | XP_028703453.1 | 1067 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |