Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260966.1 | NP_001247895.1 | 455 | 67 | ||
XM_015150590.2 | XP_015006076.1 | 961 | 67 | |||
GenBank mRNA | JU322623.1 | - | 444 | 67 | ||
JU475846.1 | - | 694 | 67 | |||
JV044734.1 | - | 455 | 67 | |||
JV601706.1 | - | 709 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |