Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194106.1 | NP_001181035.1 | 606 | 96 | ||
XM_028841782.1 | XP_028697615.1 | 617 | 96 | |||
GenBank mRNA | JU322812.1 | - | 506 | 96 | ||
JU475866.1 | - | 506 | 96 | |||
JV508990.1 | - | 509 | 96 | |||
JV653154.1 | - | 528 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |