Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116137.2 | XP_014971623.1 | 1268 | 149 | ||
XM_015116141.2 | XP_014971627.1 | 1268 | 149 | |||
XM_015116142.2 | XP_014971628.1 | 1268 | 149 | |||
XM_015116143.2 | XP_014971629.1 | 1070 | 149 | |||
XM_015116144.2 | XP_014971630.1 | 1070 | 149 | |||
XM_028834169.1 | XP_028690002.1 | 1176 | 149 | |||
XM_028834170.1 | XP_028690003.1 | 1388 | 149 | |||
XM_028834171.1 | XP_028690004.1 | 1268 | 149 | |||
XM_028834172.1 | XP_028690005.1 | 1388 | 149 | |||
XM_028834173.1 | XP_028690006.1 | 1590 | 149 | |||
XM_028834174.1 | XP_028690007.1 | 1388 | 149 | |||
GenBank mRNA | JU334748.1 | - | 1003 | 149 | ||
JV600214.1 | - | 1410 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |