Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265994.1 | NP_001252923.1 | 322 | 115 | ||
GenBank mRNA | JV047274.1 | - | 322 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |