Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266240.2 | NP_001253169.1 | 270 | 77 | ||
XM_015140893.2 | XP_014996379.1 | 1056 | 77 | |||
XM_028849274.1 | XP_028705107.1 | 1281 | 77 | |||
XM_028849275.1 | XP_028705108.1 | 3015 | 77 | |||
GenBank mRNA | JU321320.1 | - | 478 | 77 | ||
JU472890.1 | - | 198 | 77 | |||
JV047281.1 | - | 270 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |