Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133693.2 | XP_014989179.2 | 1436 | 190 | ||
XM_015133694.2 | XP_014989180.2 | 960 | 190 | |||
XM_015133695.2 | XP_014989181.2 | 1343 | 190 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |