Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092593.4 | XP_001092593.2 | 954 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |