Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150879.2 | XP_015006365.2 | 1552 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |