Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193984.2 | NP_001180913.1 | 465 | 81 | ||
GenBank mRNA | JU328176.1 | - | 352 | 81 | ||
JV047446.1 | - | 447 | 81 | |||
JV565140.1 | - | 421 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |