Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260513.1 | NP_001247442.1 | 194 | 90 | ||
GenBank mRNA | JV047447.1 | - | 194 | 90 | ||
JV636236.1 | - | 116 | 90 |