Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260513.1 | NP_001247442.1 | 1014 | 133 | ||
XM_015115593.2 | XP_014971079.1 | 998 | 133 | |||
XM_028832651.1 | XP_028688484.1 | 932 | 133 | |||
GenBank mRNA | JV047447.1 | - | 1014 | 133 | ||
JV636236.1 | - | 936 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |