Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083074.4 | XP_001083074.1 | 999 | 71 | ||
GenBank mRNA | JV065222.1 | - | 968 | 71 | ||
JV655519.1 | - | 838 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |