Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257429.1 | NP_001244358.1 | 253 | 119 | ||
XM_015124569.2 | XP_014980055.1 | 491 | 119 | |||
XM_015124577.2 | XP_014980063.1 | 286 | 119 | |||
XM_015124586.2 | XP_014980072.1 | 533 | 119 | |||
XM_028829820.1 | XP_028685653.1 | 393 | 119 | |||
XM_028829848.1 | XP_028685681.1 | 464 | 119 | |||
GenBank mRNA | JU335661.1 | - | 253 | 119 | ||
JV044794.1 | - | 254 | 119 | |||
JV567514.1 | - | 243 | 119 | |||
JV601143.1 | - | 254 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |