Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194617.2 | NP_001181546.1 | 702 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |