Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140324.2 | XP_014995810.1 | 95 | 90 | ||
XM_015140325.2 | XP_014995811.1 | 94 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |