Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140324.2 | XP_014995810.1 | 661 | 87 | ||
XM_015140325.2 | XP_014995811.1 | 660 | 87 | |||
GenBank mRNA | JV647158.1 | - | 563 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |