Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261270.1 | NP_001248199.1 | 1584 | 151 | ||
XM_015143611.2 | XP_014999097.1 | 2536 | 151 | |||
XM_015143612.2 | XP_014999098.1 | 1724 | 151 | |||
XM_015143613.2 | XP_014999099.1 | 1992 | 151 | |||
GenBank mRNA | JU322221.1 | - | 1657 | 151 | ||
JU476121.1 | - | 1584 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |