Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194084.2 | NP_001181013.1 | 1418 | 149 | ||
XM_028849183.1 | XP_028705016.1 | 1399 | 149 | |||
GenBank mRNA | JU322585.1 | - | 1356 | 149 | ||
JU472617.1 | - | 1351 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |