Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266056.1 | NP_001252985.1 | 604 | 100 | ||
GenBank mRNA | JU322099.1 | - | 604 | 100 | ||
JV549051.1 | - | 230 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |