Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193806.1 | NP_001180735.1 | 728 | 139 | ||
GenBank mRNA | JU321250.1 | - | 724 | 139 | ||
JU321251.1 | - | 724 | 139 | |||
JU476167.1 | - | 708 | 139 | |||
JU476168.1 | - | 708 | 139 | |||
JV639346.1 | - | 733 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |