Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260660.1 | NP_001247589.1 | 745 | 106 | ||
XM_028827328.1 | XP_028683161.1 | 588 | 106 | |||
GenBank mRNA | JU323198.1 | - | 745 | 106 | ||
JU473259.1 | - | 738 | 106 | |||
JV639379.1 | - | 747 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |