Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTAQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265979.1 | NP_001252908.1 | 142 | 173 | ||
GenBank mRNA | JU322754.1 | - | 142 | 173 | ||
JU478142.1 | - | 166 | 173 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |