Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083177.4 | XP_001083177.1 | 1355 | 141 | ||
GenBank mRNA | JU323499.1 | - | 1345 | 141 | ||
JU332288.1 | - | 1345 | 141 | |||
JU476214.1 | - | 1319 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |