Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM38B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194325.1 | NP_001181254.1 | 387 | 163 | ||
GenBank mRNA | JU322549.1 | - | 377 | 163 | ||
JV637314.1 | - | 385 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |