Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266952.1 | NP_001253881.1 | 1160 | 100 | ||
XM_015131361.2 | XP_014986847.1 | 1755 | 100 | |||
GenBank mRNA | JU335249.1 | - | 1156 | 100 | ||
JU472675.1 | - | 1160 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |