Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149557.2 | XP_015005043.2 | 322 | 87 | ||
XM_015149559.2 | XP_015005045.2 | 392 | 87 | |||
XM_028828148.1 | XP_028683981.1 | 457 | 87 | |||
XM_028828149.1 | XP_028683982.1 | 348 | 87 | |||
XM_028828150.1 | XP_028683983.1 | 914 | 87 | |||
XM_028828151.1 | XP_028683984.1 | 914 | 87 | |||
XM_028828152.1 | XP_028683985.1 | 914 | 87 | |||
XM_028828153.1 | XP_028683986.1 | 914 | 87 | |||
XM_028828154.1 | XP_028683987.1 | 914 | 87 | |||
XM_028828156.1 | XP_028683989.1 | 914 | 87 | |||
GenBank mRNA | JV718004.1 | - | 304 | 87 |