Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082016.4 | XP_001082016.1 | 588 | 141 | ||
GenBank mRNA | JU327322.1 | - | 320 | 141 | ||
JV718028.1 | - | 297 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |