Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127003.2 | XP_014982489.2 | 937 | 155 | ||
XM_028842169.1 | XP_028698002.1 | 937 | 155 | |||
GenBank mRNA | JU335433.1 | - | 872 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |