Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093783.4 | XP_001093783.2 | 519 | 168 | ||
XM_015149581.2 | XP_015005067.2 | 541 | 168 | |||
XM_015149582.2 | XP_015005068.2 | 512 | 168 | |||
XM_015149586.2 | XP_015005072.2 | 601 | 168 | |||
GenBank mRNA | JU335449.1 | - | 520 | 168 | ||
JU476393.1 | - | 526 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |